Learn More
Meso Scale Discovery V-PLEX Plus Human IL-1a Kit
SDP
Catalog No. 5000082198
Encompass
The Meso Scale Discovery V-PLEX Human IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 1 alpha (IL-1 ) in cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence to provide high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL
Catalog No. 50-000-82198 Supplier Meso Scale Discovery Supplier No. K151RBG2
The Meso Scale Discovery V-PLEX Human IL-1 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 1 alpha (IL-1 ) in cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence to provide high sensitivity a wide dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 1500 pg/mL