Learn More
Meso Scale Discovery V-PLEX Plus Human IL-1SY Kit
SDP
Catalog No. 5000079182
Encompass
The V-PLEX Plus Human IL-1 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and other biological samples The assay uses a plate that has been pre-coated with capture antibodies and utilizes a V-PLEX Human IL-1 Detection Kit for detection The kit includes a 96-well plate detection antibodies read buffer calibrator diluent wash buffer and a vial of lyophilized SULFO-TAG labeled detection antibody The assay has high sensitivity and a broad dynamic range from 0 06-1500 pg/mL
Catalog No. 50-000-79182 Supplier Meso Scale Discovery Supplier No. K151QPG1
The V-PLEX Plus Human IL-1 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and other biological samples The assay uses a plate that has been pre-coated with capture antibodies and utilizes a V-PLEX Human IL-1 Detection Kit for detection The kit includes a 96-well plate detection antibodies read buffer calibrator diluent wash buffer and a vial of lyophilized SULFO-TAG labeled detection antibody The assay has high sensitivity and a broad dynamic range from 0 06-1500 pg/mL