Learn More
Meso Scale Discovery V-PLEX Plus Human IL-1SY Kit
SDP
Catalog No. 5000079183
Encompass
The V-PLEX Plus Human IL-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 1500 pg/mL
Catalog No. 50-000-79183 Supplier Meso Scale Discovery Supplier No. K151QPG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human IL-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human interleukin 1 beta (IL-1 ) in cell culture supernatants serum plasma and other biological fluids The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 1500 pg/mL