Learn More
Meso Scale Discovery V-PLEX Plus Human IL-2 Kit
SDP
Catalog No. 5000078582
Encompass
The V-PLEX Plus Human IL-2 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 2 (IL-2) in cell culture supernatants serum plasma and other biological samples The kit includes a 96-well plate coated with a monoclonal antibody to human IL-2 a detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 06 pg/mL and a broad dynamic range from 0 06-1500 pg/mL Sample incubation time is 2 hours
Catalog No. 50-000-78582 Supplier Meso Scale Discovery Supplier No. K151QQG2
The V-PLEX Plus Human IL-2 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 2 (IL-2) in cell culture supernatants serum plasma and other biological samples The kit includes a 96-well plate coated with a monoclonal antibody to human IL-2 a detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 06 pg/mL and a broad dynamic range from 0 06-1500 pg/mL Sample incubation time is 2 hours