Learn More
Meso Scale Discovery V-PLEX Plus Human IL-2 Kit
SDP
Catalog No. 5000079839
Encompass
The V-PLEX Plus Human IL-2 Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) to measure human interleukin 2 (IL-2) in various samples It includes a 96-well plate with a monoclonal antibody for IL-2 a detection antibody with an ECL label calibrators and buffers The assay s sensitivity is 0 06 pg/mL with a range of 0 06-1500 pg/mL requiring 50 L of sample and providing results in under 3 hours using an MSD ECL instrument It s suitable for immunology and inflammation research
Catalog No. 50-000-79839 Supplier Meso Scale Discovery Supplier No. K151QQG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human IL-2 Kit from Meso Scale Discovery is an immunoassay using electrochemiluminescence (ECL) to measure human interleukin 2 (IL-2) in various samples It includes a 96-well plate with a monoclonal antibody for IL-2 a detection antibody with an ECL label calibrators and buffers The assay s sensitivity is 0 06 pg/mL with a range of 0 06-1500 pg/mL requiring 50 L of sample and providing results in under 3 hours using an MSD ECL instrument It s suitable for immunology and inflammation research