Learn More
Meso Scale Discovery V-PLEX Plus Human IL-3 Kit
SDP
Catalog No. 5000079751
Encompass
The Meso Scale Discovery Human IL-3 Singleplex Assay Kit is a sensitive immunoassay using electrochemiluminescence (ECL) to measure human interleukin 3 (IL-3) in various samples It includes a plate with capture and detection antibodies calibrator and buffers The detection range is 0 3-2500 pg/mL requiring 50 L or less sample volume The ECL platform offers faster results wider dynamic range and higher sensitivity than ELISA Suitable for research and clinical applications measuring IL-3 cytokine levels
Catalog No. 50-000-79751 Supplier Meso Scale Discovery Supplier No. K151WZG2
The Meso Scale Discovery Human IL-3 Singleplex Assay Kit is a sensitive immunoassay using electrochemiluminescence (ECL) to measure human interleukin 3 (IL-3) in various samples It includes a plate with capture and detection antibodies calibrator and buffers The detection range is 0 3-2500 pg/mL requiring 50 L or less sample volume The ECL platform offers faster results wider dynamic range and higher sensitivity than ELISA Suitable for research and clinical applications measuring IL-3 cytokine levels