Learn More
Meso Scale Discovery V-PLEX Plus Human IL-3 Kit
SDP
Catalog No. 5000081839
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-3 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 3 (IL-3) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and includes reagents for testing 39 samples in duplicate The electrochemiluminescence is measured using an MSD instrument such as SECTOR Imager QuickPlex or MESO QuickPlex SQ 120
Catalog No. 50-000-81839 Supplier Meso Scale Discovery Supplier No. K151WZG1
The Meso Scale Discovery V-PLEX Plus Human IL-3 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 3 (IL-3) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and includes reagents for testing 39 samples in duplicate The electrochemiluminescence is measured using an MSD instrument such as SECTOR Imager QuickPlex or MESO QuickPlex SQ 120