Learn More
Meso Scale Discovery V-PLEX Plus Human IL-4 Kit
SDP
Catalog No. 5000078792
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-4 Kit is an immunoassay for measuring human interleukin 4 (IL-4) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD s SECTOR Imager systems aiding in immunology and inflammation research
Catalog No. 50-000-78792 Supplier Meso Scale Discovery Supplier No. K151QRG1
The Meso Scale Discovery V-PLEX Plus Human IL-4 Kit is an immunoassay for measuring human interleukin 4 (IL-4) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD s SECTOR Imager systems aiding in immunology and inflammation research