Learn More
Meso Scale Discovery V-PLEX Plus Human IL-4 Kit
SDP
Catalog No. 5000081676
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-4 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 4 (IL-4) in various biological fluids It includes a 96-well plate with a monoclonal antibody for IL-4 a detection antibody with an electrochemiluminescent label calibrators and buffers The assay s sensitivity is 0 3 pg/mL with a range of 0 3-2500 pg/mL Using MSD s SULFO-TAG labels and MESO QuickPlex SQ 120 it offers accurate IL-4 quantitation in 3 5 hours
Catalog No. 50-000-81676 Supplier Meso Scale Discovery Supplier No. K151QRG2
The Meso Scale Discovery V-PLEX Plus Human IL-4 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 4 (IL-4) in various biological fluids It includes a 96-well plate with a monoclonal antibody for IL-4 a detection antibody with an electrochemiluminescent label calibrators and buffers The assay s sensitivity is 0 3 pg/mL with a range of 0 3-2500 pg/mL Using MSD s SULFO-TAG labels and MESO QuickPlex SQ 120 it offers accurate IL-4 quantitation in 3 5 hours