Learn More
Meso Scale Discovery V-PLEX Plus Human IL-5 Kit
SDP
Catalog No. 5000078581
Encompass
The Meso Scale Discovery V-PLEX Human IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 5 (IL-5) in cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence to provide high sensitivity broad dynamic range and rapid results The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions to generate a standard curve The assay has high sensitivity with a lower limit of detection of 0 04 pg/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-78581 Supplier Meso Scale Discovery Supplier No. K151QSG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-5 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 5 (IL-5) in cell culture supernatants serum plasma and other biological samples The assay utilizes MSD s proprietary V-PLEX technology based on electrochemiluminescence to provide high sensitivity broad dynamic range and rapid results The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions to generate a standard curve The assay has high sensitivity with a lower limit of detection of 0 04 pg/mL and an upper limit of 10 000 pg/mL