Learn More
Meso Scale Discovery V-PLEX Plus Human IL-5 Kit
SDP
Catalog No. 5000079181
Encompass
The Meso Scale Discovery V-PLEX Human IL-5 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 5 (IL-5) in various biological fluids It uses MSD s V-PLEX technology for high sensitivity wide dynamic range and quick results The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators for a standard curve It detects IL-5 from 0 04 to 1500 pg/mL with a total assay time of 3 5 hours
Catalog No. 50-000-79181 Supplier Meso Scale Discovery Supplier No. K151QSG2
The Meso Scale Discovery V-PLEX Human IL-5 Kit is an electrochemiluminescent immunoassay for measuring human interleukin 5 (IL-5) in various biological fluids It uses MSD s V-PLEX technology for high sensitivity wide dynamic range and quick results The kit includes a plate with capture antibodies detection antibodies with an electrochemiluminescent label and calibrators for a standard curve It detects IL-5 from 0 04 to 1500 pg/mL with a total assay time of 3 5 hours