Learn More
Meso Scale Discovery V-PLEX Plus Human IL-6 Kit
SDP
Catalog No. 5000078795
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a capture antibody specific for human IL-6 a detection antibody conjugated to an electrochemiluminescent label calibrators diluents wash buffers and other reagents required to perform the assay The kit has a detection range of 0 06-1500 pg/mL and exhibits no cross-reactivity with other cytokines The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-78795 Supplier Meso Scale Discovery Supplier No. K151QXG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a capture antibody specific for human IL-6 a detection antibody conjugated to an electrochemiluminescent label calibrators diluents wash buffers and other reagents required to perform the assay The kit has a detection range of 0 06-1500 pg/mL and exhibits no cross-reactivity with other cytokines The assay takes approximately 3 5 hours to complete