Learn More
Meso Scale Discovery V-PLEX Plus Human IL-6 Kit
SDP
Catalog No. 5000079185
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a capture antibody specific for human IL-6 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-2500 pg/mL The total assay time is 3 5 hours This kit allows for the accurate and sensitive quantification of human IL-6 levels
Catalog No. 50-000-79185 Supplier Meso Scale Discovery Supplier No. K151QXG2
The Meso Scale Discovery V-PLEX Plus Human IL-6 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a capture antibody specific for human IL-6 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-2500 pg/mL The total assay time is 3 5 hours This kit allows for the accurate and sensitive quantification of human IL-6 levels