Learn More
Meso Scale Discovery V-PLEX Plus Human IL-7 Kit
SDP
Catalog No. 5000079884
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with a capture antibody specific for human IL-7 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 11 pg/mL and a 7-log dynamic range from 0 49 pg/mL to 10 000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantitation of human IL-7 levels in biological samples
Catalog No. 50-000-79884 Supplier Meso Scale Discovery Supplier No. K151RCG1
The Meso Scale Discovery V-PLEX Plus Human IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in cell culture supernatants serum and plasma The kit contains a 96-well plate coated with a capture antibody specific for human IL-7 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 11 pg/mL and a 7-log dynamic range from 0 49 pg/mL to 10 000 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantitation of human IL-7 levels in biological samples