Learn More
Meso Scale Discovery V-PLEX Plus Human IL-7 Kit
SDP
Catalog No. 5000081679
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody specific for human IL-7 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 10 pg/mL and a broad dynamic range from 0 53 - 1500 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantification of human IL-7
Catalog No. 50-000-81679 Supplier Meso Scale Discovery Supplier No. K151RCG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human IL-7 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody specific for human IL-7 a detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 10 pg/mL and a broad dynamic range from 0 53 - 1500 pg/mL The total assay time is 3 5 hours This kit allows for the accurate quantification of human IL-7