Learn More
Meso Scale Discovery V-PLEX Plus Human IL-7 Kit
SDP
Catalog No. 5000082199
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-7 Kit is a 5-plate assay kit for the quantitative measurement of human interleukin 7 (IL-7) in cell culture supernatants serum plasma and other biological fluids The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity wide dynamic range and low sample consumption The assay has high specificity for human IL-7 with no cross-reactivity to other cytokines Each kit provides reagents for 5 x 96 tests in a quick 4 5 hour protocol The kit can accurately quantitate human IL-7 over a range of 3 2-10 000 pg/mL
Catalog No. 50-000-82199 Supplier Meso Scale Discovery Supplier No. K151RCG2
The Meso Scale Discovery V-PLEX Plus Human IL-7 Kit is a 5-plate assay kit for the quantitative measurement of human interleukin 7 (IL-7) in cell culture supernatants serum plasma and other biological fluids The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity wide dynamic range and low sample consumption The assay has high specificity for human IL-7 with no cross-reactivity to other cytokines Each kit provides reagents for 5 x 96 tests in a quick 4 5 hour protocol The kit can accurately quantitate human IL-7 over a range of 3 2-10 000 pg/mL