Learn More
Meso Scale Discovery V-PLEX Plus Human IL-8 Kit
SDP
Catalog No. 5000078838
Encompass
The V-PLEX Plus Human IL-8 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 8 (IL-8) in cell culture supernatants serum plasma and other biological samples The kit includes a 96-well plate coated with a monoclonal antibody to human IL-8 a detection antibody conjugated to an ECL label calibrators and reagents required for the assay The sensitivity of the assay is 0 3 pg/mL with an assay range from 0 3-2500 pg/mL The kit takes approximately 3 5 hours to run and can measure up to 80 samples in duplicate
Catalog No. 50-000-78838 Supplier Meso Scale Discovery Supplier No. K151RAG1
The V-PLEX Plus Human IL-8 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 8 (IL-8) in cell culture supernatants serum plasma and other biological samples The kit includes a 96-well plate coated with a monoclonal antibody to human IL-8 a detection antibody conjugated to an ECL label calibrators and reagents required for the assay The sensitivity of the assay is 0 3 pg/mL with an assay range from 0 3-2500 pg/mL The kit takes approximately 3 5 hours to run and can measure up to 80 samples in duplicate