Learn More
Meso Scale Discovery V-PLEX Plus Human IL-8 Kit
SDP
Catalog No. 5000080590
Encompass
The Meso Scale Discovery V-PLEX Plus Human IL-8 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 8 (IL-8) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to complete This highly sensitive and reproducible kit allows for the accurate quantification of IL-8 levels
Catalog No. 50-000-80590 Supplier Meso Scale Discovery Supplier No. K151RAG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human IL-8 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 8 (IL-8) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 0 06-2500 pg/mL and takes approximately 3 5 hours to complete This highly sensitive and reproducible kit allows for the accurate quantification of IL-8 levels