Learn More
Meso Scale Discovery V-PLEX Plus Human IL-8 Kit
SDP
Catalog No. 5000081268
Encompass
The V-PLEX Plus Human IL-8 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 8 (IL-8) in cell culture supernatants serum plasma and other biological samples The kit includes a 96-well plate coated with a monoclonal antibody to human IL-8 a detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 09 pg/mL and an upper limit of quantification of 1500 pg/mL It can detect native and recombinant human IL-8 with a total assay time of 3 5 hours
Catalog No. 50-000-81268 Supplier Meso Scale Discovery Supplier No. K151RAG2
The V-PLEX Plus Human IL-8 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 8 (IL-8) in cell culture supernatants serum plasma and other biological samples The kit includes a 96-well plate coated with a monoclonal antibody to human IL-8 a detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 09 pg/mL and an upper limit of quantification of 1500 pg/mL It can detect native and recombinant human IL-8 with a total assay time of 3 5 hours