Learn More
Meso Scale Discovery V-PLEX Plus Human IL-9 Kit
SDP
Catalog No. 5000081797
Encompass
Meso Scale Discovery s V-PLEX Plus Cytokine and Chemokine Panel is a multiplex immunoassay measuring up to 45 human cytokines and chemokines from one sample Using electrochemiluminescence detection it offers high sensitivity and a broad dynamic range The panel includes cytokines like IL-6 TNF-alpha IFN-gamma and chemokines like IP-10 MCP-1 MIP-1b Requiring just 12 5 L of sample it works with serum plasma culture supernatants and other samples providing quantitative results in 3 5 hours efficiently profiling inflammation and immune responses
Catalog No. 50-000-81797 Supplier Meso Scale Discovery Supplier No. K151XCG2
Meso Scale Discovery s V-PLEX Plus Cytokine and Chemokine Panel is a multiplex immunoassay measuring up to 45 human cytokines and chemokines from one sample Using electrochemiluminescence detection it offers high sensitivity and a broad dynamic range The panel includes cytokines like IL-6 TNF-alpha IFN-gamma and chemokines like IP-10 MCP-1 MIP-1b Requiring just 12 5 L of sample it works with serum plasma culture supernatants and other samples providing quantitative results in 3 5 hours efficiently profiling inflammation and immune responses