Learn More
Meso Scale Discovery V-PLEX Plus Human Insulin Kit
SDP
Catalog No. 5000079165
Encompass
The Meso Scale Discovery V-PLEX Plus Human Insulin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human insulin in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a range of 3 13-10 000 pg/mL and takes approximately 3 5 hours to complete It uses an electrochemiluminescent detection method to provide high sensitivity and wide dynamic range while only requiring a small sample volume of 25 L
Catalog No. 50-000-79165 Supplier Meso Scale Discovery Supplier No. K151S5G1
The Meso Scale Discovery V-PLEX Plus Human Insulin Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human insulin in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a range of 3 13-10 000 pg/mL and takes approximately 3 5 hours to complete It uses an electrochemiluminescent detection method to provide high sensitivity and wide dynamic range while only requiring a small sample volume of 25 L