Learn More
Meso Scale Discovery V-PLEX Plus Human Insulin Kit
SDP
Catalog No. 5000081684
Encompass
The V-PLEX Plus Human Insulin Kit from Meso Scale Discovery is an immunoassay for measuring human insulin in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results comparable to ELISA The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents With a dynamic range of 0 05-1000 pg/mL and high sensitivity it measures insulin levels in about 3 5 hours
Catalog No. 50-000-81684 Supplier Meso Scale Discovery Supplier No. K151S5G2
Add to Cart
Add to Cart
The V-PLEX Plus Human Insulin Kit from Meso Scale Discovery is an immunoassay for measuring human insulin in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results comparable to ELISA The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents With a dynamic range of 0 05-1000 pg/mL and high sensitivity it measures insulin levels in about 3 5 hours