Learn More
Meso Scale Discovery V-PLEX Plus Human Leptin Kit
SDP
Catalog No. 5000078301
Encompass
The V-PLEX Plus Human Leptin Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of leptin in human serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2000 pg/mL for leptin
Catalog No. 50-000-78301 Supplier Meso Scale Discovery Supplier No. K151V5G2
The V-PLEX Plus Human Leptin Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of leptin in human serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator diluent The assay has a typical sensitivity of 0 04 pg/mL and an upper limit of quantification of 2000 pg/mL for leptin