Learn More
Meso Scale Discovery V-PLEX Plus Human Leptin Kit
SDP
Catalog No. 5000080374
Encompass
The Meso Scale Discovery V-PLEX Plus Human Leptin Kit is a highly sensitive immunoassay that quantitatively measures human leptin in serum plasma and culture supernatants The kit uses electrochemiluminescence detection technology and provides accurate reproducible results in just 3 5 hours It requires a small sample volume of 50 L and has a lower limit of detection of 26 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized recombinant human leptin standard
Catalog No. 50-000-80374 Supplier Meso Scale Discovery Supplier No. K151V5G1
The Meso Scale Discovery V-PLEX Plus Human Leptin Kit is a highly sensitive immunoassay that quantitatively measures human leptin in serum plasma and culture supernatants The kit uses electrochemiluminescence detection technology and provides accurate reproducible results in just 3 5 hours It requires a small sample volume of 50 L and has a lower limit of detection of 26 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized recombinant human leptin standard