Learn More
Meso Scale Discovery V-PLEX Plus Human MCP-1 Kit
SDP
Catalog No. 5000079270
Encompass
The V-PLEX Plus Human MCP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human MCP-1 in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against human MCP-1 detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 2 4-10 000 pg/mL and exhibits minimal cross-reactivity with other analytes It allows for high-sensitivity quantification of MCP-1 levels from small sample volumes
Catalog No. 50-000-79270 Supplier Meso Scale Discovery Supplier No. K151NNG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human MCP-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human MCP-1 in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies against human MCP-1 detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 2 4-10 000 pg/mL and exhibits minimal cross-reactivity with other analytes It allows for high-sensitivity quantification of MCP-1 levels from small sample volumes