Learn More
Meso Scale Discovery V-PLEX Plus Human MCP-4 Kit
SDP
Catalog No. 5000078654
Encompass
The V-PLEX Plus Human MCP-4 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 4 (MCP-4) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized Calibrator and Controls to allow construction of a standard curve and ensure assay performance The electrochemiluminescent detection method allows for high sensitivity and a broad dynamic range while only requiring a small sample volume The kit can quantify human MCP-4 concentrations between 0 06 - 1500 pg/mL
Catalog No. 50-000-78654 Supplier Meso Scale Discovery Supplier No. K151NOG1
The V-PLEX Plus Human MCP-4 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 4 (MCP-4) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Calibrator Diluent and lyophilized Calibrator and Controls to allow construction of a standard curve and ensure assay performance The electrochemiluminescent detection method allows for high sensitivity and a broad dynamic range while only requiring a small sample volume The kit can quantify human MCP-4 concentrations between 0 06 - 1500 pg/mL