Learn More
Meso Scale Discovery V-PLEX Plus Human MCP-4 Kit
SDP
Catalog No. 5000079265
Encompass
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit measures 30 human cytokines and chemokines in serum plasma and culture supernatants using electrochemiluminescence detection It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators controls buffers diluent and seals It efficiently profiles cytokines and chemokines related to inflammation immunity angiogenesis and hematopoiesis
Catalog No. 50-000-79265 Supplier Meso Scale Discovery Supplier No. K151NOG2
The Meso Scale Discovery V-PLEX Plus Human Cytokine 30-Plex Kit measures 30 human cytokines and chemokines in serum plasma and culture supernatants using electrochemiluminescence detection It offers high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies detection antibodies with MSD SULFO-TAG calibrators controls buffers diluent and seals It efficiently profiles cytokines and chemokines related to inflammation immunity angiogenesis and hematopoiesis