Learn More
Meso Scale Discovery V-PLEX Plus Human MDC Kit
SDP
Catalog No. 5000078651
Encompass
The V-PLEX Plus Human MDC Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of 10 human cytokines and chemokines (IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IFN- TNF- IP-10 and MCP-1) from cell culture supernatants serum plasma and other biological fluids The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and vials for the preparation of standard curves
Catalog No. 50-000-78651 Supplier Meso Scale Discovery Supplier No. K151NPG2
The V-PLEX Plus Human MDC Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of 10 human cytokines and chemokines (IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IFN- TNF- IP-10 and MCP-1) from cell culture supernatants serum plasma and other biological fluids The kit utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and vials for the preparation of standard curves