Learn More
Meso Scale Discovery V-PLEX Plus Human MDC Kit
SDP
Catalog No. 5000080766
Encompass
The V-PLEX Plus Human MDC Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring 10 human cytokines and chemokines (IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IFN- TNF- IP-10 MCP-1) in cell culture supernatants serum plasma and other samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibodies buffers diluent calibrators and controls for rapid quantification of immune mediators
Catalog No. 50-000-80766 Supplier Meso Scale Discovery Supplier No. K151NPG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human MDC Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring 10 human cytokines and chemokines (IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IFN- TNF- IP-10 MCP-1) in cell culture supernatants serum plasma and other samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume requirements The kit includes a 96-well plate with capture antibodies detection antibodies buffers diluent calibrators and controls for rapid quantification of immune mediators