Learn More
Meso Scale Discovery V-PLEX Plus Human MIP-1a Kit
SDP
Catalog No. 5000078650
Encompass
The Meso Scale Discovery V-PLEX Plus Human MIP-1a Kit is a multiplex immunoassay for measuring human MIP-1a in various biological fluids It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use The kit includes a 96-well plate with capture antibodies labeled detection antibodies calibrators diluent wash buffer read buffer and plate seals It shows high specificity with under 0 3% cross-reactivity The kit allows up to 80 tests in duplicate with a testing time of 3 5 hours
Catalog No. 50-000-78650 Supplier Meso Scale Discovery Supplier No. K151NQG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human MIP-1a Kit is a multiplex immunoassay for measuring human MIP-1a in various biological fluids It uses electrochemiluminescence detection offering high sensitivity a 5-log dynamic range and low sample use The kit includes a 96-well plate with capture antibodies labeled detection antibodies calibrators diluent wash buffer read buffer and plate seals It shows high specificity with under 0 3% cross-reactivity The kit allows up to 80 tests in duplicate with a testing time of 3 5 hours