Learn More
Meso Scale Discovery V-PLEX Plus Human MIP-1a Kit
SDP
Catalog No. 5000079264
Encompass
The V-PLEX Human MIP-1 alpha Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of human MIP-1 alpha in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate seals The electrochemiluminescent assay has a detection range of 1 53-2500 pg/mL and requires 25 L of sample The total assay time is approximately 3 5 hours This highly sensitive and reproducible kit can help researchers study chemokine biology and inflammation
Catalog No. 50-000-79264 Supplier Meso Scale Discovery Supplier No. K151NQG1
The V-PLEX Human MIP-1 alpha Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of human MIP-1 alpha in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate seals The electrochemiluminescent assay has a detection range of 1 53-2500 pg/mL and requires 25 L of sample The total assay time is approximately 3 5 hours This highly sensitive and reproducible kit can help researchers study chemokine biology and inflammation