Learn More
Meso Scale Discovery V-PLEX Plus Human MIP-1SY Kit
SDP
Catalog No. 5000079129
Encompass
The Meso Scale Discovery K151NRG-1 assay kit is a 96-well plate kit for the quantitative measurement of human Neuregulin-1 in cell culture supernatants serum plasma and other biological samples The kit uses MSD s electrochemiluminescence technology and provides high sensitivity a broad dynamic range and requires a low sample volume The assay time is just 3 5 hours and the kit includes a plate detection antibody calibrator and assay diluent The kit has a sensitivity of 0 3 pg/mL and an upper limit of quantitation of 1500 pg/mL
Catalog No. 50-000-79129 Supplier Meso Scale Discovery Supplier No. K151NRG1
The Meso Scale Discovery K151NRG-1 assay kit is a 96-well plate kit for the quantitative measurement of human Neuregulin-1 in cell culture supernatants serum plasma and other biological samples The kit uses MSD s electrochemiluminescence technology and provides high sensitivity a broad dynamic range and requires a low sample volume The assay time is just 3 5 hours and the kit includes a plate detection antibody calibrator and assay diluent The kit has a sensitivity of 0 3 pg/mL and an upper limit of quantitation of 1500 pg/mL