Learn More
Meso Scale Discovery V-PLEX Plus Human MIP-1SY Kit
SDP
Catalog No. 5000079926
Encompass
Meso Scale Discovery s V-PLEX Plus Cytokine Panel 1 (human) kit enables the quantitative measurement of 10 cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from various samples This electrochemiluminescent multiplex immunoassay uses MSD s MULTI-ARRAY technology suitable for biomarker discovery and research The kit includes a 96-well plate antibodies calibrators controls diluent read buffer and necessary reagents Results are analyzed on MSD s SECTOR Imager for high sensitivity and dynamic range
Catalog No. 50-000-79926 Supplier Meso Scale Discovery Supplier No. K151NRG2
Meso Scale Discovery s V-PLEX Plus Cytokine Panel 1 (human) kit enables the quantitative measurement of 10 cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from various samples This electrochemiluminescent multiplex immunoassay uses MSD s MULTI-ARRAY technology suitable for biomarker discovery and research The kit includes a 96-well plate antibodies calibrators controls diluent read buffer and necessary reagents Results are analyzed on MSD s SECTOR Imager for high sensitivity and dynamic range