Learn More
Meso Scale Discovery V-PLEX Plus Human PP Kit
SDP
Catalog No. 5000079820
Encompass
The V-PLEX Human PP Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of Pancreatic Polypeptide in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate sealers The electrochemiluminescent immunoassay has a detection range of 3 2-10 000 pg/mL and requires 25 L sample size The assay has high sensitivity requires no wash steps and results can be read on the MSD instrument platforms This kit allows for the accurate quantification of Pancreatic Polypeptide levels in human samples
Catalog No. 50-000-79820 Supplier Meso Scale Discovery Supplier No. K151Y5G1
The V-PLEX Human PP Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of Pancreatic Polypeptide in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate sealers The electrochemiluminescent immunoassay has a detection range of 3 2-10 000 pg/mL and requires 25 L sample size The assay has high sensitivity requires no wash steps and results can be read on the MSD instrument platforms This kit allows for the accurate quantification of Pancreatic Polypeptide levels in human samples