Learn More
Meso Scale Discovery V-PLEX Plus Human PP Kit
SDP
Catalog No. 5000080109
Encompass
The V-PLEX Plus Human PP Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of Properdin in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate sealers The electrochemiluminescent assay has a detection range of 78-5000 pg/mL and requires 25 L of sample The total assay time is 3 5 hours The kit includes reagents for 96 tests and has a 12 month shelf life when stored at 2-8 C
Catalog No. 50-000-80109 Supplier Meso Scale Discovery Supplier No. K151Y5G2
The V-PLEX Plus Human PP Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of Properdin in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies standards buffers substrate and plate sealers The electrochemiluminescent assay has a detection range of 78-5000 pg/mL and requires 25 L of sample The total assay time is 3 5 hours The kit includes reagents for 96 tests and has a 12 month shelf life when stored at 2-8 C