Learn More
Meso Scale Discovery V-PLEX Plus Human SAA Kit
SDP
Catalog No. 5000081300
Encompass
The V-PLEX Plus Human SAA Kit from Meso Scale Discovery is an immunoassay kit for the quantitative measurement of serum amyloid A (SAA) in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibody detection antibody calibrator diluent read buffer wash buffer and plate seals The electrochemiluminescent assay has a detection range of 0 3-1000 ng/mL SAA with high sensitivity and specificity The kit takes 3 5 hours to run and is designed for use on Meso Scale Discovery instruments
Catalog No. 50-000-81300 Supplier Meso Scale Discovery Supplier No. K151SSG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human SAA Kit from Meso Scale Discovery is an immunoassay kit for the quantitative measurement of serum amyloid A (SAA) in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibody detection antibody calibrator diluent read buffer wash buffer and plate seals The electrochemiluminescent assay has a detection range of 0 3-1000 ng/mL SAA with high sensitivity and specificity The kit takes 3 5 hours to run and is designed for use on Meso Scale Discovery instruments