Learn More
Meso Scale Discovery V-PLEX Plus Human SAA Kit
SDP
Catalog No. 5000081707
Encompass
The V-PLEX Plus Human SAA Kit from Meso Scale Diagnostics is an immunoassay that quantitatively measures serum amyloid A (SAA) in human serum plasma and culture supernatant samples The kit utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 1500 pg/mL
Catalog No. 50-000-81707 Supplier Meso Scale Discovery Supplier No. K151SSG2
The V-PLEX Plus Human SAA Kit from Meso Scale Diagnostics is an immunoassay that quantitatively measures serum amyloid A (SAA) in human serum plasma and culture supernatant samples The kit utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 0 3 pg/mL and an upper limit of quantification of 1500 pg/mL