Learn More
Meso Scale Discovery V-PLEX Plus Human TARC Kit
SDP
Catalog No. 5000078652
Encompass
The V-PLEX Plus Human TARC Kit from Meso Scale Discovery is a multiplex immunoassay measuring Th2-associated chemokine receptor (TARC/CCL17) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for simultaneous TARC detection with high sensitivity and broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody Read Buffer Calibrator Blend and Sample Diluent TARC is measured in pg/mL with a detection limit of 0 3 pg/mL aiding in understanding Th2-mediated inflammation and immune responses
Catalog No. 50-000-78652 Supplier Meso Scale Discovery Supplier No. K151NTG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Human TARC Kit from Meso Scale Discovery is a multiplex immunoassay measuring Th2-associated chemokine receptor (TARC/CCL17) in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for simultaneous TARC detection with high sensitivity and broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody Read Buffer Calibrator Blend and Sample Diluent TARC is measured in pg/mL with a detection limit of 0 3 pg/mL aiding in understanding Th2-mediated inflammation and immune responses