Learn More
Meso Scale Discovery V-PLEX Plus Human Tie-2 Kit
SDP
Catalog No. 5000080846
Encompass
The Meso Scale Discovery V-PLEX Plus Human TIE-2 Kit is a multiplex immunoassay that measures human TIE-2 levels in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology allowing simultaneous measurement of TIE-2 and other biomarkers of interest The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay Results are analyzed on MSD s imaging platform and data analysis software This quantitative assay provides sensitive and reproducible results with a broad dynamic range
Catalog No. 50-000-80846 Supplier Meso Scale Discovery Supplier No. K151KWG2
The Meso Scale Discovery V-PLEX Plus Human TIE-2 Kit is a multiplex immunoassay that measures human TIE-2 levels in serum plasma and cell culture supernatants This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology allowing simultaneous measurement of TIE-2 and other biomarkers of interest The kit includes a 96-well plate coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay Results are analyzed on MSD s imaging platform and data analysis software This quantitative assay provides sensitive and reproducible results with a broad dynamic range