Learn More
Meso Scale Discovery V-PLEX Plus Human TSLP Kit
SDP
Catalog No. 5000081420
Encompass
The V-PLEX Plus Human TSLP Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Thymic Stromal Lymphopoietin (TSLP) across multiple samples using electrochemiluminescence (ECL) detection technology The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound and calibrator solutions for generating a standard curve This assay has high sensitivity wide dynamic range and requires low sample volume The kit can test up to 80 samples in duplicate and has a testing time of 3 5 hours
Catalog No. 50-000-81420 Supplier Meso Scale Discovery Supplier No. K151XEG1
The V-PLEX Plus Human TSLP Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of human Thymic Stromal Lymphopoietin (TSLP) across multiple samples using electrochemiluminescence (ECL) detection technology The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL compound and calibrator solutions for generating a standard curve This assay has high sensitivity wide dynamic range and requires low sample volume The kit can test up to 80 samples in duplicate and has a testing time of 3 5 hours