Learn More
Meso Scale Discovery V-PLEX Plus Human TSLP Kit
SDP
Catalog No. 5000081994
Encompass
The V-PLEX Plus Human TSLP Kit from Meso Scale Discovery is a 96-well plate assay that measures human thymic stromal lymphopoietin (TSLP) in cell culture supernatants serum plasma and other biological samples The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for natural and recombinant human TSLP with no cross-reactivity to related cytokines The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator The assay can quantify TSLP concentrations from 15 6 to 1000 pg/mL
Catalog No. 50-000-81994 Supplier Meso Scale Discovery Supplier No. K151XEG2
The V-PLEX Plus Human TSLP Kit from Meso Scale Discovery is a 96-well plate assay that measures human thymic stromal lymphopoietin (TSLP) in cell culture supernatants serum plasma and other biological samples The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The assay has high specificity for natural and recombinant human TSLP with no cross-reactivity to related cytokines The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents and calibrator The assay can quantify TSLP concentrations from 15 6 to 1000 pg/mL