Learn More
Meso Scale Discovery V-PLEX Plus Human VCAM-1 Kit
SDP
Catalog No. 5000080541
Encompass
The V-PLEX Plus Human VCAM-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of human VCAM-1 in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The V-PLEX kit can reliably quantify VCAM-1 across a range of 0 53 - 2500 pg/mL This product would be useful for studying inflammatory biomarkers and endothelial activation
Catalog No. 50-000-80541 Supplier Meso Scale Discovery Supplier No. K151SRG2
The V-PLEX Plus Human VCAM-1 Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the quantitative measurement of human VCAM-1 in serum plasma and cell culture supernatants The kit utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volume The V-PLEX kit can reliably quantify VCAM-1 across a range of 0 53 - 2500 pg/mL This product would be useful for studying inflammatory biomarkers and endothelial activation