Learn More
Meso Scale Discovery V-PLEX Plus Human VEGF-C Kit
SDP
Catalog No. 5000079292
Encompass
The V-PLEX Plus Human VEGF-C Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for quantifying human VEGF-C in various biological fluids It includes a 96-well plate with a VEGF-C capture antibody a detection antibody with an electrochemiluminescent label and calibrators and controls for a standard curve The assay detects 3 2-10 000 pg/mL and shows no cross-reactivity with other VEGF isoforms offering high sensitivity and a wide dynamic range for accurate VEGF-C quantification
Catalog No. 50-000-79292 Supplier Meso Scale Discovery Supplier No. K151LTG2
The V-PLEX Plus Human VEGF-C Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for quantifying human VEGF-C in various biological fluids It includes a 96-well plate with a VEGF-C capture antibody a detection antibody with an electrochemiluminescent label and calibrators and controls for a standard curve The assay detects 3 2-10 000 pg/mL and shows no cross-reactivity with other VEGF isoforms offering high sensitivity and a wide dynamic range for accurate VEGF-C quantification