Learn More
Meso Scale Discovery V-PLEX Plus Human VEGF-C Kit
SDP
Catalog No. 5000079552
Encompass
The V-PLEX Plus Human VEGF-C Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human VEGF-C in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a VEGF-C capture antibody a VEGF-C detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 31 pg/mL and a range of 0 31-2500 pg/mL Sample incubation time is 2 hours The ECL signal produced is proportional to the amount of VEGF-C present in the sample
Catalog No. 50-000-79552 Supplier Meso Scale Discovery Supplier No. K151LTG1
The V-PLEX Plus Human VEGF-C Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human VEGF-C in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a VEGF-C capture antibody a VEGF-C detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay has a sensitivity of 0 31 pg/mL and a range of 0 31-2500 pg/mL Sample incubation time is 2 hours The ECL signal produced is proportional to the amount of VEGF-C present in the sample