Learn More
Meso Scale Discovery V-PLEX Plus Human VEGF-D Kit
SDP
Catalog No. 5000079553
Encompass
The Meso Scale Discovery V-PLEX Plus Human VEGF-D Kit is an electrochemiluminescent immunoassay for measuring human VEGF-D in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive seals The assay sensitivity is 1 3 pg/mL with a range of 1 3-1000 pg/mL using MSD s SECTOR Imager for detection and takes about 3 5 hours The kit tests 40 samples in duplicate
Catalog No. 50-000-79553 Supplier Meso Scale Discovery Supplier No. K151LUG1
The Meso Scale Discovery V-PLEX Plus Human VEGF-D Kit is an electrochemiluminescent immunoassay for measuring human VEGF-D in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent wash buffer read buffer and adhesive seals The assay sensitivity is 1 3 pg/mL with a range of 1 3-1000 pg/mL using MSD s SECTOR Imager for detection and takes about 3 5 hours The kit tests 40 samples in duplicate