Learn More
Meso Scale Discovery V-PLEX Plus Human VEGF Kit
SDP
Catalog No. 5000078511
Encompass
The V-PLEX Plus Human VEGF Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human VEGF in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals Sensitivity is 0 31 pg/mL with an upper quantification limit of 2500 pg/mL The assay takes about 3 5 hours and is used for studying angiogenesis vascular permeability and VEGF signaling
Catalog No. 50-000-78511 Supplier Meso Scale Discovery Supplier No. K151RHG2
The V-PLEX Plus Human VEGF Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human VEGF in various biological fluids It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals Sensitivity is 0 31 pg/mL with an upper quantification limit of 2500 pg/mL The assay takes about 3 5 hours and is used for studying angiogenesis vascular permeability and VEGF signaling