Learn More
Meso Scale Discovery V-PLEX Plus Human VEGF Kit
SDP
Catalog No. 5000081293
Encompass
The Meso Scale Discovery V-PLEX Plus Human VEGF Kit is a multiplex immunoassay that measures vascular endothelial growth factor (VEGF) in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to enable the simultaneous detection of VEGF on a QuickPlex SQ 120 instrument The kit includes a VEGF-specific antibody coated plate detection antibody calibrator and assay diluent optimized to provide sensitive and reproducible quantitation of human VEGF levels with a wide dynamic range This kit allows high-throughput analysis of VEGF from small sample volumes
Catalog No. 50-000-81293 Supplier Meso Scale Discovery Supplier No. K151RHG4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human VEGF Kit is a multiplex immunoassay that measures vascular endothelial growth factor (VEGF) in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to enable the simultaneous detection of VEGF on a QuickPlex SQ 120 instrument The kit includes a VEGF-specific antibody coated plate detection antibody calibrator and assay diluent optimized to provide sensitive and reproducible quantitation of human VEGF levels with a wide dynamic range This kit allows high-throughput analysis of VEGF from small sample volumes