Learn More
Meso Scale Discovery V-PLEX Plus Mouse IFN-y Kit
SDP
Catalog No. 5000081331
Encompass
The V-PLEX Plus Mouse IFN-gamma Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of mouse interferon gamma (IFN-) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibodies against mouse IFN- detection antibodies labeled with an electrochemiluminescent compound calibrator solutions diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-10 000 pg/mL It utilizes MSD s proprietary QuickPlex SQ 120 instrument and provides rapid results in 3 5 hours
Catalog No. 50-000-81331 Supplier Meso Scale Discovery Supplier No. K152QOG4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Plus Mouse IFN-gamma Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of mouse interferon gamma (IFN-) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with capture antibodies against mouse IFN- detection antibodies labeled with an electrochemiluminescent compound calibrator solutions diluent wash buffer read buffer and adhesive plate seals The assay has a sensitivity of 0 3 pg/mL and a broad dynamic range from 0 3-10 000 pg/mL It utilizes MSD s proprietary QuickPlex SQ 120 instrument and provides rapid results in 3 5 hours