Learn More
Meso Scale Discovery V-PLEX Plus Mouse IFN-y Kit
SDP
Catalog No. 5000081513
Encompass
The V-PLEX Plus Mouse IFN-gamma Kit from Meso Scale Discovery is a multiplex immunoassay for measuring mouse IFN- in various samples It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent wash buffer read buffer and seals The assay s sensitivity is 0 3 pg/mL with a range of 0 3-10 000 pg/mL It uses MSD s SECTOR Imager for detection ensuring high sensitivity and a wide range with small sample volumes The kit provides all necessary reagents for mouse sample analysis
Catalog No. 50-000-81513 Supplier Meso Scale Discovery Supplier No. K152QOG1
The V-PLEX Plus Mouse IFN-gamma Kit from Meso Scale Discovery is a multiplex immunoassay for measuring mouse IFN- in various samples It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators diluent wash buffer read buffer and seals The assay s sensitivity is 0 3 pg/mL with a range of 0 3-10 000 pg/mL It uses MSD s SECTOR Imager for detection ensuring high sensitivity and a wide range with small sample volumes The kit provides all necessary reagents for mouse sample analysis