Learn More
Meso Scale Discovery V-PLEX Plus Mouse IL-10 Kit
SDP
Catalog No. 5000078625
Encompass
The Meso Scale Discovery V-PLEX Plus Mouse IL-10 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 10 (IL-10) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MESO QuickPlex SQ 120 for plate reading The kit includes reagents for one 96-well plate and can assay 40 samples in duplicate
Catalog No. 50-000-78625 Supplier Meso Scale Discovery Supplier No. K152QUG2
The Meso Scale Discovery V-PLEX Plus Mouse IL-10 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse interleukin 10 (IL-10) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MESO QuickPlex SQ 120 for plate reading The kit includes reagents for one 96-well plate and can assay 40 samples in duplicate